missing translation for 'onlineSavingsMsg'
Learn More

Lithium tri-tert-butoxyaluminohydride, 93-98%

Product Code. 11995791 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11995791

Brand: Thermo Scientific Acros 125165000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 17476-04-9 | C12H28AlLiO3 | 254.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 17476-04-9
Molecular Formula C12H28AlLiO3
Molecular Weight (g/mol) 254.27
MDL Number MFCD00011532

Spécification

Melting Point 319.0°C
Density 0.9
Infrared Spectrum Authentic
Assay Percent Range 93% min
Packaging Glass Bottle packed in tin can
Linear Formula LiAI[OC(CH3)3]3H
Quantity 500 g
Fieser 01,620; 02,251; 03,188; 04,312; 10,248; 12,286; 13,342; 16,197
Solubility Information Solubility in water: reacts violently. Other solubilities: 36g/100g thf (25°C),2g/100g diethyl ether (25°C),41g/100g diglyme (25°C)
Sublimation Point 280 °C
Formula Weight 254.27
Percent Purity 93 to 98%
Physical Form Powder
Chemical Name or Material Lithium tri-tert-butoxyaluminohydride, 93-98%
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • Lithium tri-tert-butoxyaluminohydride
Mention d'avertissement
  • Danger
Catégorie de danger
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Mention de danger
  • H314-Causes severe skin burns and eye damage.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis