missing translation for 'onlineSavingsMsg'
Learn More
Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals Product Code.: 11319488

Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 11319488
10 mL
Click to view available options
Quantity:
10 mL
50 mL
This item is not returnable. View return policy

Product Code. 11319488

Brand: Thermo Scientific Alfa Aesar 043324.AB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17476-04-9
Molecular Formula C12H28AlLiO3
MDL Number MFCD00011532
Synonym Lithium hydridotris(tert-butoxy)aluminate

Specifications

Color Colorless
Density 0.904 g/mL
Flash Point −17°C (1°F)
Packaging ChemSeal™ bottles
Linear Formula Li[(CH3)3CO]3AlH
Quantity 10 mL
UN Number UN3399
Sensitivity Moisture sensitive
Solubility Information It is soluble in organic solvents at low temperatures
Formula Weight 254.28
Physical Form Liquid
Chemical Name or Material Lithium tri-tert-butoxyaluminum hydride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.