missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium triethylborohydride, 1.7M solution in THF, Thermo Scientific Chemicals
£53.00 - £310.66
Chemical Identifiers
CAS | 22560-16-3 |
---|---|
Molecular Formula | C6H16BLi |
Molecular Weight (g/mol) | 105.94 |
MDL Number | MFCD00011703 |
InChI Key | WCJAYABJWDIZAJ-UHFFFAOYSA-N |
IUPAC Name | lithium(1+) triethylboranuide |
SMILES | [Li+].CC[BH-](CC)CC |
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
22560-16-3 | |
105.94 | |
WCJAYABJWDIZAJ-UHFFFAOYSA-N | |
[Li+].CC[BH-](CC)CC |
C6H16BLi | |
MFCD00011703 | |
lithium(1+) triethylboranuide |
Especificaciones
22560-16-3 | |
LiB(C2H5)3H | |
0.8900g/mL | |
WCJAYABJWDIZAJ-UHFFFAOYSA-N | |
lithium(1+) triethylboranuide | |
105.94 | |
−17°C |
C6H16BLi | |
MFCD00011703 | |
Solubility in water: reacts. | |
[Li+].CC[BH-](CC)CC | |
105.94 | |
0.89 | |
Lithium triethylborohydride |
Seguridad y manipulación
Nombre del producto
Palabra de advertencia
Clasificación
Indicaciones de peligro
Consejos de prudencia
- Lithium triethylborohydride, 1.7M solution in THF
Palabra de advertencia
- Danger
Clasificación
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Pyrophoric liquid Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Indicaciones de peligro
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Consejos de prudencia
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only