Learn More
Lithium trifluoroacetate monohydrate, 97%, Thermo Scientific Chemicals
£41.30 - £157.00
Chemical Identifiers
CAS | 2923-17-3 |
---|---|
Molecular Formula | C2F3LiO2 |
Molecular Weight (g/mol) | 119.96 |
MDL Number | MFCD00013216,MFCD00149663 |
InChI Key | HSFDLPWPRRSVSM-UHFFFAOYSA-M |
Synonym | lithium trifluoroacetate, acetic acid, trifluoro-, lithium salt, lithium 2,2,2-trifluoroacetate, lithiumtrifluoroacetate, acmc-1cfal, lithotab trifluoroacetate, acetic acid, 2,2,2-trifluoro-, lithium salt 1:1, lithium trifluoroacetate, monohydrate, lithium 1+ trifluoroacetate, lithio 2,2,2-trifluoroacetate |
PubChem CID | 23661853 |
IUPAC Name | lithium;2,2,2-trifluoroacetate |
SMILES | [Li+].[O-]C(=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11369349
|
Thermo Scientific Alfa Aesar
088938.09 |
10 g |
£41.30
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359349
|
Thermo Scientific Alfa Aesar
088938.18 |
50 g |
£157.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2923-17-3 | |
119.96 | |
HSFDLPWPRRSVSM-UHFFFAOYSA-M | |
23661853 | |
[Li+].[O-]C(=O)C(F)(F)F |
C2F3LiO2 | |
MFCD00013216,MFCD00149663 | |
lithium trifluoroacetate, acetic acid, trifluoro-, lithium salt, lithium 2,2,2-trifluoroacetate, lithiumtrifluoroacetate, acmc-1cfal, lithotab trifluoroacetate, acetic acid, 2,2,2-trifluoro-, lithium salt 1:1, lithium trifluoroacetate, monohydrate, lithium 1+ trifluoroacetate, lithio 2,2,2-trifluoroacetate | |
lithium;2,2,2-trifluoroacetate |
Specifications
2923-17-3 | |
C2F3LiO2 | |
MFCD00013216,MFCD00149663 | |
Hygroscopic | |
HSFDLPWPRRSVSM-UHFFFAOYSA-M | |
lithium;2,2,2-trifluoroacetate | |
23661853 | |
97% | |
Lithium trifluoroacetate monohydrate |
White | |
LiCO2CF3·H2O | |
UN2811 | |
lithium trifluoroacetate, acetic acid, trifluoro-, lithium salt, lithium 2,2,2-trifluoroacetate, lithiumtrifluoroacetate, acmc-1cfal, lithotab trifluoroacetate, acetic acid, 2,2,2-trifluoro-, lithium salt 1:1, lithium trifluoroacetate, monohydrate, lithium 1+ trifluoroacetate, lithio 2,2,2-trifluoroacetate | |
[Li+].[O-]C(=O)C(F)(F)F | |
119.96 | |
137.97 (119.96 Anhydrous) | |
Powder |
Safety and Handling
GHS H Statement
H300
Fatal if swallowed.
P264b-P270-P301+P310-P330-P501c
H300
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 220-878-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only