missing translation for 'onlineSavingsMsg'
Learn More

Lithium (trimethylsilyl)acetylide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

£162.81 - £954.75

Chemical Identifiers

CAS 54655-07-1, 109-99-9
Molecular Formula C5H9LiSi
Molecular Weight (g/mol) 104.15
MDL Number MFCD00075059
InChI Key WDDOQHLJFOUQMW-UHFFFAOYSA-N
Synonym trimethylsilyl ethynyllithium, lithium, trimethylsilyl ethynyl, lithium 1+ ion trimethylsilyl ethyne, lithium trimethylsilylacetylide, lithio trimethylsilyl acetylene, lithium trimethylsilyl acetylide, lithium ethynyl trimethyl silane, lithium trimethylsilylacetylenide, zvxxeonxfwsciz-uhfffaoysa-n
PubChem CID 3431600
IUPAC Name lithium;ethynyl(trimethyl)silane
SMILES [Li+].C[Si](C)(C)C#[C-]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10359213
View Documents
Thermo Scientific Acros
380651000
100 mL AcroSeal™ Glass Bottle in Tin Can
£162.81
100mL
Please to purchase this item. Need a web account? Register with us today!
10310524
View Documents
Thermo Scientific Acros
380658000
800 mL AcroSeal™ Glass Bottle
£954.75
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54655-07-1, 109-99-9
104.15
WDDOQHLJFOUQMW-UHFFFAOYSA-N
3431600
[Li+].C[Si](C)(C)C#[C-]
C5H9LiSi
MFCD00075059
trimethylsilyl ethynyllithium, lithium, trimethylsilyl ethynyl, lithium 1+ ion trimethylsilyl ethyne, lithium trimethylsilylacetylide, lithio trimethylsilyl acetylene, lithium trimethylsilyl acetylide, lithium ethynyl trimethyl silane, lithium trimethylsilylacetylenide, zvxxeonxfwsciz-uhfffaoysa-n
lithium;ethynyl(trimethyl)silane

Specifications

54655-07-1, 109-99-9
Solution
MFCD00075059
WDDOQHLJFOUQMW-UHFFFAOYSA-N
lithium;ethynyl(trimethyl)silane
3431600
0.4 to 0.6M
Lithium (trimethylsilyl)acetylide
Yellow to Orange
C5H9LiSi
trimethylsilyl ethynyllithium, lithium, trimethylsilyl ethynyl, lithium 1+ ion trimethylsilyl ethyne, lithium trimethylsilylacetylide, lithio trimethylsilyl acetylene, lithium trimethylsilyl acetylide, lithium ethynyl trimethyl silane, lithium trimethylsilylacetylenide, zvxxeonxfwsciz-uhfffaoysa-n
[Li+].C[Si](C)(C)C#[C-]
104.15
104.15
0.8800g/mL
Videos
Safety and Handling

Safety and Handling

GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for se

Danger

Product Identifier
  • Lithium (trimethylsilyl)acetylide, 0.5M solution in THF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only