missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium vanadium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Lithium vanadium oxide is used in developing polymer lithium ion (PLI) batteries and also in coatings, nanowires, nanofibers, plastics, and textiles In alloy and catalyst applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 250 g |
UN Number | UN3285 |
Solubility Information | Soluble in water |
Formula Weight | 105.88 |
Percent Purity | 99.9% |
Physical Form | Powder |
Assay | (metals basis) |
Chemical Name or Material | Lithium vanadium oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'