missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Litmus, Indicator (pH 5-8), for titration, Honeywell Fluka™
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Packaging:
Glass Bottle
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Especificaciones
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000% |
Linear Formula | CHCl3 |
Grade | Analytical Reagent |
Packaging | Glass Bottle |
Quantity | 25 g |
Chemical Name or Material | Litmus, Indicator (pH 5-8), for titration |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'