missing translation for 'onlineSavingsMsg'
Learn More

Litmus, Indicator (pH 5-8), for titration, Honeywell Fluka™

Indicator (pH 5-8), for titration

£273.74 - £3261.01

Chemical Identifiers

CAS 1393-92-6
MDL Number MFCD00000826
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15696030
View Documents
Honeywell-Fluka
61280-25G
25 g Glass Bottle
£273.74
25g
Estimated Shipment: 01-04-2025
to see stock.
Add to basket
 
15606040
View Documents
Honeywell-Fluka
61280-500G
500 g Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1393-92-6
MFCD00000826

Specifications

1393-92-6
≤100.0000%
MFCD00000826
Litmus, Indicator (pH 5-8), for titration
≥90.0000%
CHCl3
Analytical Reagent
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1620 days from date of manufacture

missing translation for 'einecsNumber' : 215-739-6

Recommended Storage : Room Temp

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.