missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit, for UV excitation
The LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.
£144.00 - £382.00
Specifications
Cell Permeability | Impermeant |
---|---|
Cell Type | Eukaryotic Cells |
Color | Blue |
Description | LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit, for UV excitation |
Detection Method | Fluorescence |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10114812
|
Molecular Probes™
L23105 |
200 Assays |
£369.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15582241
|
Molecular Probes™
L34961 |
80 Assays |
£144.00
1 set |
In Stock
Log in to see stock. |
|||||
15585313
|
Molecular Probes™
L34962 |
400 Assays |
£382.00
1 set |
In Stock
Log in to see stock. |
|||||
Description
- Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
- Kit has been optimized and validated for use with a UV laser flow cytometer
- Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
- Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
- If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
- LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs
Optimal brightness
- The LIVE/DEAD™ Fixable Blue Stain was selected based on its fluorescent properties to provide a bright signal when excited with a UV laser
- The blue-fluorescent reactive dye has an excitation maximum of ∼350 nm, making it ideal for use with an UV laser, and an emission of ∼450 nm
- By switching viability staining from a channel off of the blue 488 nm laser to the UV laser, a channel is freed up on the flow cytometer for other reagents that are difficult to find in colors other than FITC or PE
- By using the UV laser, there is no spectral overlap issues with other common dyes, thus eliminating the need to compensate the viability stains from other stains in the panel
- Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, it is an ideal choice for multi-color experiments
Specifications
Impermeant | |
Blue | |
Fluorescence | |
Flow Cytometer | |
LIVE/DEAD™ Fixable Blue Dead Cell Stain | |
350 nm | |
Tube(s) | |
Room Temperature |
Eukaryotic Cells | |
LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit, for UV excitation | |
Viability Assay | |
Dead Cell Stain Kit | |
450 nm | |
Solid | |
LIVE/DEAD™ | |
DMSO (Dimethylsulfoxide) |
For Research Use Only. Not for use in diagnostic procedures.