missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE/DEAD™ Fixable Lime (506) Viability Kit, for 405 nm excitation

The LIVE/DEAD Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry.

Brand:  Invitrogen™ L34989

 View more versions of this product

Product Code. 17344553

  • £144.00 / Pack of 80

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry
  • Cells labeled with LIVE/DEAD Fixable viability dyes can be washed, fixed, permeabilized, and stained for intracellular antigens while maintaining distinct live and dead cell populations
  • The narrow and unique emission spectra are ideal for expanding high-parameter flow cytometry experiments
  • Designed to provide a bright signal with minimal overlap into adjacent channels, simplifying spectral compensation
  • Since live and dead cells can be discriminated using a single flow cytometer channel, these viability dyes are an ideal choice for multi-color experiments.
  • Their unique spectral fingerprints are a valuable addition to spectral flow cytometry experiments
  • Dyes have been conveniently packaged in single-use vials to help ensure dye stability and performance over time
  • Dissolve the dye in the included anhydrous DMSO and use immediately, or store at -20°C, avoiding freeze-thaw cycles
  • The LIVE/DEAD Fixable Dead Cell Stain Sampler Kit provides one vial (40 assays) of each of eight available dead cell stains
TRUSTED_SUSTAINABILITY
Specifications

Specifications

80 Assays
Fluorescence
Cell Viability Kit
Tube(s)
Contains 2 vials of LIVE/DEAD fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
405/506 nm
DMSO (Dimethylsulfoxide)
LIVE/DEAD™
LIVE/DEAD™ Fixable Lime (506) Viability Kit, for 405 nm excitation
Flow Cytometry
LIVE/DEAD Fixable Lime (506) Cell Stain
Lime
Eukaryotic Cells
405 nm
Room Temperature
Flow Cytometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.