missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIVE/DEAD™ Fixable Near IR (876) Viability Kit, for 808 nm excitation
The LIVE/DEAD Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry.
£138.00 - £377.00
Specifications
Detection Method | Fluorescence |
---|---|
Conjugate | LIVE/DEAD Fixable Near IR (876) Cell Stain |
For Use With (Application) | Flow Cytometry |
Product Type | Cell Viability Kit |
Description | LIVE/DEAD™ Fixable Near IR (876) Viability Kit, for 808 nm excitation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17497772
|
Invitrogen™
L34980 |
80 Assays |
£144.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17408262
|
Invitrogen™
L34980 |
80 Assays | N/A | ||||||
17418262
|
Invitrogen™
L34981 |
200 Assays |
£327.00
Pack of 200 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17428262
|
Invitrogen™
L34982 |
400 Assays |
£377.00
Pack of 400 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry
- Cells labeled with LIVE/DEAD Fixable viability dyes can be washed, fixed, permeabilized, and stained for intracellular antigens while maintaining distinct live and dead cell populations
- The narrow and unique emission spectra are ideal for expanding high-parameter flow cytometry experiments
- Designed to provide a bright signal with minimal overlap into adjacent channels, simplifying spectral compensation
- Since live and dead cells can be discriminated using a single flow cytometer channel, these viability dyes are an ideal choice for multi-color experiments.
- Their unique spectral fingerprints are a valuable addition to spectral flow cytometry experiments
- Dyes have been conveniently packaged in single-use vials to help ensure dye stability and performance over time
- Dissolve the dye in the included anhydrous DMSO and use immediately, or store at -20°C, avoiding freeze-thaw cycles
- The LIVE/DEAD Fixable Dead Cell Stain Sampler Kit provides one vial (40 assays) of each of eight available dead cell stains
Specifications
Fluorescence | |
Flow Cytometry | |
LIVE/DEAD™ Fixable Near IR (876) Viability Kit, for 808 nm excitation | |
Near-IR | |
Eukaryotic Cells | |
808 nm | |
Room Temperature | |
Flow Cytometer |
LIVE/DEAD Fixable Near IR (876) Cell Stain | |
Cell Viability Kit | |
Tubes | |
Viability Assay | |
849/876 nm | |
DMSO (Dimethylsulfoxide) | |
LIVE/DEAD™ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title