missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit, for 633 or 635 nm excitation
Determines the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation.
£152.00 - £392.00
Specifications
Cell Permeability | Impermeant |
---|---|
Cell Type | Eukaryotic Cells |
Color | Near-IR |
Description | LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit, for 633 or 635 nm excitation |
Detection Method | Fluorescence |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15519340
|
Invitrogen™
L34975 |
80 assays |
£152.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10154363
|
Invitrogen™
L10119 |
200 Assays |
£289.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15550745
|
Invitrogen™
L34976 |
400 Assays |
£392.00
400 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
- Kit has been optimized and validated for use with a red laser flow cytometer
- Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
- Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
- If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
- LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs
Optimal brightness
- The LIVE/DEAD™ Fixable Near-IR stain was selected based on its fluorescent properties to provide a bright signal when excited with a red laser
- The near-IR fluorescent reactive dye has an excitation maximum of ∼633 nm making it ideal for use with the red or HeNe laser with an emission of ∼750 nm
- Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, it is an ideal choice for multi-color experiments
Specifications
Impermeant | |
Near-IR | |
Fluorescence | |
Flow Cytometer | |
LIVE/DEAD™ FixableNear-IR Dead Cell Stain | |
Solid | |
LIVE/DEAD™ | |
DMSO (Dimethylsulfoxide) |
Eukaryotic Cells | |
LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit, for 633 or 635 nm excitation | |
Viability Assay | |
Stain | |
633 nm | |
Tube(s) | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit, for 633 or 635 nm excitation