missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE/DEAD™ Fixable Olive (557) Viability Kit, for 488 nm excitation

The LIVE/DEAD Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry.

£144.00 - £377.00

Specifications

Detection Method Fluorescence
Conjugate LIVE/DEAD Fixable Olive (557) Dead Cell Stain
For Use With (Application) Flow Cytometry
Description LIVE/DEAD™ Fixable Olive (557) Viability Kit, for 488 nm excitation
Format Tubes
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17478252
View Documents
Invitrogen™
L34977
80 Assays
£144.00
Pack of 80
Please to purchase this item. Need a web account? Register with us today!
17488252
View Documents
Invitrogen™
L34978
200 Assays
£327.00
Pack of 200
Please to purchase this item. Need a web account? Register with us today!
17498252
View Documents
Invitrogen™
L34979
400 Assays
£377.00
Pack of 400
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry
  • Cells labeled with LIVE/DEAD Fixable viability dyes can be washed, fixed, permeabilized, and stained for intracellular antigens while maintaining distinct live and dead cell populations
  • The narrow and unique emission spectra are ideal for expanding high-parameter flow cytometry experiments
  • Designed to provide a bright signal with minimal overlap into adjacent channels, simplifying spectral compensation
  • Since live and dead cells can be discriminated using a single flow cytometer channel, these viability dyes are an ideal choice for multi-color experiments.
  • Their unique spectral fingerprints are a valuable addition to spectral flow cytometry experiments
  • Dyes have been conveniently packaged in single-use vials to help ensure dye stability and performance over time
  • Dissolve the dye in the included anhydrous DMSO and use immediately, or store at -20°C, avoiding freeze-thaw cycles
  • The LIVE/DEAD Fixable Dead Cell Stain Sampler Kit provides one vial (40 assays) of each of eight available dead cell stains
Specifications

Specifications

Fluorescence
Flow Cytometry
Tubes
Eukaryotic Cells
488 nm
Room Temperature
Flow Cytometer
LIVE/DEAD Fixable Olive (557) Dead Cell Stain
LIVE/DEAD™ Fixable Olive (557) Viability Kit, for 488 nm excitation
Olive
479/557 nm
DMSO (Dimethylsulfoxide)
LIVE/DEAD™
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.