missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ LIVE/DEAD™ Fixable Violet Dead Cell Stain Kit, for 405 nm excitation

The LIVE/DEAD™ Fixable Violet Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.

Brand:  Molecular Probes™ L34955

324.64 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 10645203

  • £367.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
  • Kit has been optimized and validated for use with a violet laser flow cytometer
  • Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
  • Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
  • If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
  • LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs

Low compensation

  • The LIVE/DEAD™ Fixable Violet Stain was selected based on its fluorescent properties to minimize compensation between other violet dyes and dyes that excite off of the 488 nm blue laser
  • The violet-fluorescent reactive dye has an excitation maximum of ∼416 nm and is excited well with the 405 nm violet laser
  • It has an emission maxima of ∼451 nm, so it can be collected in the first channel on most violet laser flow cytometers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Impermeant
Violet
LIVE/DEAD™ Fixable Violet Dead Cell Stain Kit, for 405 nm excitation
Viability Assay
Stain
451 nm
Solid
LIVE/DEAD™
Room Temperature
Eukaryotic Cells
Contains 5 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Fluorescence
Flow Cytometer
LIVE/DEAD™ Fixable Violet Dead Cell Stain
416 nm
Tube(s)
200 Assays
DMSO (Dimethylsulfoxide)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.