missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ LIVE/DEAD™ BacLight™ Bacterial Viability and Counting Kit, for flow cytometry

The LIVE/DEAD™ BacLight™ Bacterial Viability and Counting Kit allows researchers to reliably distinguish and quantitate live and dead bacteria with the aid of a flow cytometer, even in a mixed population containing a range of bacterial types.

Brand:  Molecular Probes™ L34856

526.69 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.


Product Code. 10503673

  • £790.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

The LIVE/DEAD™ BacLight™ Bacterial Viability and Counting Kit allows researchers to reliably distinguish and quantitate live and dead bacteria with the aid of a flow cytometer, even in a mixed population containing a range of bacterial types. This kit uses a mixture of two nucleic acid stains - green-fluorescent SYTO™ 9 dye and red-fluorescent propidium iodide - for viability determinations, and a calibrated suspension of microspheres for accurate sample volume measurements.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 1 vial SYTO™ 9 nucleic acid stain (200 μL, 3.34 mM in DMSO), 1 vial of propidium iodide (200 μL, 20 mM in DMSO), and 1 vial of a microsphere standard (1.0 mL of 6.0 μm diameter microspheres).

Store in refrigerator (2–8°C) and protect from light.
Tube
SYTO™ 9, propidium iodide
100 reactions kit
SYTO™ 9: 485/498, PI: 535/617
Fluorescence
Bacterial Viability and Counting Kit
BacLight™, LIVE/DEAD™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3096426 Certificate TypeCertificate of Analysis Date22/12/2024 Catalog NumberL34856
Lot Number2800323 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog NumberL34856
Lot Number2585787 Certificate TypeCertificate of Analysis Date05/05/2023 Catalog NumberL34856
Lot Number2351982 Certificate TypeCertificate of Analysis Date19/07/2021 Catalog NumberL34856
Lot Number2261454 Certificate TypeCertificate of Analysis Date12/11/2020 Catalog NumberL34856
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ LIVE/DEAD™ BacLight™ Bacterial Viability and Counting Kit, for flow cytometry > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.