Learn More
Molecular Probes™ LIVE/DEAD™ FungaLight™ Yeast Viability Kit, for flow cytometry
The LIVE/DEAD FungaLight Yeast Viability Kit enables researchers to easily, reliably, and quantitatively distinguish live and dead yeast in minutes by flow cytometry.
Brand: Molecular Probes™ L34952
761.90 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
The LIVE/DEAD™ FungaLight™ Yeast Viability Kit enables researchers to easily, reliably and quantitatively distinguish live and dead yeast in minutes by flow cytometry. The kit contains solutions of the SYTO 9 green-fluorescent nucleic acid stain and the red-fluorescent nucleic acid stain, propidium iodide. These stains differ both in their spectral characteristics and in their ability to penetrate healthy yeast cells. When used alone, the SYTO 9 stain generally labels all yeast in a population - those with intact membranes and those with damaged membranes. In contrast, propidium iodide penetrates only yeast with damaged membranes, causing a reduction in the SYTO 9 stain fluorescence by fluorescence resonance energy transfer (FRET) when both dyes are present. As a result, yeast with intact membranes stain fluorescent green, whereas yeast with damaged membranes stain fluorescent red.
Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Impermeant, Permeant | |
Contains 1 vial of SYTO™ 9 dye (solution in DMSO), and 1 vial of propidium iodide, 300 μL (solution in DMSO). Store in freezer (-5°C to -30°C) and protect from light. |
|
Fluorescence | |
Flow Cytometer | |
SYTO™ 9, propidium iodide | |
485/535 nm | |
Tube(s) | |
200 Assays kit |
Green, Red | |
LIVE/DEAD™ FungaLight™ Yeast Viability Kit, for flow cytometry | |
Viability Assay | |
Yeast Viability Kit | |
498/617 nm | |
Solution | |
FungaLight™, LIVE/DEAD™ | |
Room Temperature |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.