Learn More
Abnova™ LMO7 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FS0061.100uL
This item is not returnable.
View return policy
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
FBX20,FBXO20,KIAA0858,LOMP | |
4008 | |
100 μL |
Tube | |
LIM domain 7 | |
LMO7 | |
DNA/RNA | |
Liquid |