missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMOD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Bio-Techne NBP1-85898
This item is not returnable.
View return policy
Description
LMOD3 Polyclonal specifically detects LMOD3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LMOD3 | |
Polyclonal | |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Q0VAK6 | |
LMOD3 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VTDKAFKEQRDRPEAQEQSEKKISKLDPKKLALDTSFLKVSTRPSGNQTDLDGSLRRVRKNDPDMKELNLNNIENIPKEMLLDFVNAMKK | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DKFZp313F0135, leiomodin 3 (fetal), Leiomodin, fetal form, leiomodin-3 | |
Rabbit | |
Affinity Purified | |
RUO | |
56203 | |
Human | |
IgG |
For Research Use Only