Learn More
Thermo Scientific™ LN2 Level Monitor
Product Code. 10164623 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Locator 4 Plus
Locator 8 Plus
Locator Jr. Plus
Voltage:
100/240 V
230 V
Unit Size:
Each
Product Code. 10164623
Brand: Thermo Scientific™ CN509X7
Only null left
Add to basket
Add to basket
Safeguard irreplaceable biological specimens with the LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available.
Safeguard irreplaceable biological specimens with the Thermo Scientific™ LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available. Designed to minimize liquid nitrogen evaporation, the monitor is for use with Thermo Scientific™ Locator™ and Locator Plus cryogenic storage systems.
Ultrasonic Level Monitor
  • Minimizes conduction and liquid nitrogen evaporation
  • Easy-to-read indicator continuously displays liquid nitrogen volume in 1/8 increments
  • Audible and visual alarms signal low level liquid nitrogen conditions
TRUSTED_SUSTAINABILITY
Specifications
Description Level Monitor
Type LN2 Measuring Device
For Use With (Equipment) Locator 4 Plus
Monitoring Options Alarm
Voltage 230 V