Learn More
Thermo Scientific™ LN2 Phase Separator
Use this LN2 Phase Separator with LN2 Supply Tanks.
Brand: Thermo Scientific™ 4000571
This item is not returnable.
View return policy
Description
Use Thermo Scientific™ LN2 Phase Separator with LN2 Supply Tanks.
Specifications
LN2 phase separator, 1.3 x 3 in. (3.3 x 7.6cm) | |
LN2 Phase Separator | |
CryoPlus 1,2,3,4 |