Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points Traceable Calibration
LO-tox™ lab thermometer complete with calibration certificate.
£118.00 - £136.22
Product Code | Brand | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Price | Quantity & Availability | ||
17823008
|
Brannan™
44/814/0 + 49/101/0 |
-10°C to +330°C | 2°C | Partial Immersion | 30.5 cm |
£121.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17873028
|
Brannan™
44/802/9 + 49/101/0 |
-20°C to +110°C | 0.5°C | Partial Immersion | 30.5 cm |
£118.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17862988
|
Brannan™
44/824/0 + 49/101/0 |
-1°C to 101°C | 0.2°C | Partial Immersion | 40.5 cm |
£132.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17833028
|
Brannan™
44/808/8 + 49/101/0 |
-10°C to +50°C | 0.5°C | Total Immersion | 30.5 cm |
£119.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17883028
|
Brannan™
44/804/8 + 49/101/0 |
-10°C to +260°C | 1°C | Partial Immersion | 40.5 cm |
£119.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17822998
|
Brannan™
44/830/7 + 49/101/0 |
-10°C to +110°C | 1°C | Total Immersion | 15.5 cm |
£118.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17842998
|
Brannan™
44/819/0 + 49/101/0 |
-10°C to +360°C | 2°C | Total Immersion | 30.5 cm |
£121.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17863018
|
Brannan™
44/820/0 + 49/101/0 |
-1°C to 51°C | 0.1°C | Partial Immersion | 40.5 cm |
£131.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17843028
|
Brannan™
44/805/8 + 49/101/0 |
-10°C to +210°C | 1°C | Partial Immersion | 40.5 cm |
£119.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17892988
|
Brannan™
44/825/0 + 49/101/0 |
-1°C to 101°C | 0.2°C | Total Immersion | 40.5 cm |
£136.22
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17853018
|
Brannan™
44/801/8 + 49/101/0 |
-35°C to +50°C | 1°C | Partial Immersion | 30.5 cm |
£119.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17843038
|
Brannan™
44/810/9 + 49/101/0 |
-20°C to +110°C | 0.5°C | Total Immersion | 30.5 cm |
£118.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17823068
|
Brannan™
44/823/0 + 49/101/0 |
-1°C to 51°C | 0.2°C | Total Immersion | 40.5 cm |
£131.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17803088
|
Brannan™
44/815/0 + 49/101/0 |
-10°C to +300°C | 1°C | Partial Immersion | 30.5 cm |
£121.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17853108
|
Brannan™
44/802/8 + 49/101/0 |
-20°C to +110°C | 1°C | Partial Immersion | 30.5 cm |
£118.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17893118
|
Brannan™
44/812/8 + 49/101/0 |
-10°C to +210°C | 1°C | Total Immersion | 40.5 cm |
£120.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17813088
|
Brannan™
44/803/8 + 49/101/0 |
-20°C to +150°C | 1°C | Partial Immersion | 30.5 cm |
£122.81
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17873108
|
Brannan™
44/810/8 + 49/101/0 |
-20°C to +110°C | 1°C | Total Immersion | 30.5 cm |
£118.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17893108
|
Brannan™
44/822/0 + 49/101/0 |
-1°C to 51°C | 0.2°C | Partial Immersion | 40.5 cm |
£131.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17853088
|
Brannan™
44/811/8 + 49/101/0 |
-20°C to +150°C | 1°C | Total Immersion | 30.5 cm |
£119.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17833108
|
Brannan™
44/800/8 + 49/101/0 |
-10°C to +50°C | 0.5°C | Partial Immersion | 30.5 cm |
£119.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17863068
|
Brannan™
44/821/0 + 49/101/0 |
-1°C to 51°C | 0.1°C | Total Immersion | 40.5 cm |
£131.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17833078
|
Brannan™
44/850/7 + 49/101/0 |
-10°C to +50°C | 1°C | Total Immersion | 15.5 cm |
£118.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17823078
|
Brannan™
44/809/8 + 49/101/0 |
-35°C to +50°C | 0.5°C | Total Immersion | 30.5 cm |
£119.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Specifications
Glass | |
1.2 cm |