Learn More
Astell Scientific™ Load Sensed Process Timing
Click to view available options
For Use With (Equipment):
Astell Scientific Autoclaves
Unit Size:
Each
Description
- Optional for any Astell autoclave.
- Particularly recommended for fluid loads, e.g., culture media, where a precise sterilization temperature is likely to be critical.
- Waste (discard) loads will generally benefit from the inclusion of this option, to ensure that all parts of the load are reaching the necessary temperature.
- Allows the sterilization cycle to be controlled via the temperature achieved in the center of the load.
- When sterilizing discard, the probe should be placed in a suitable container in the center of the load or, if specialist discard containers are used, (and this is preferable), there is often a custom-designed compartment within the container to accept the probe, and prevent it from adhering to plastic waste etc.
- Sterilization period begins once the probe reaches the programmed threshold temperature.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Outside | 5-6 mm |
| For Use With (Equipment) | Astell Scientific Autoclaves |
| Product Type | Load Sensed Process Timing |