Learn More
Astell Scientific™ Load Support Plate
Support shelf to expand autoclave capacity. Astell Scientific™ Load Support Plate accommodates additional Duran bottles without the need for baskets.
£220.00
Specifications
Product Type | Load Support Plate |
---|---|
Material | Stainless Steel |
Quantity | 1 Ea. |
Description
- Expands autoclave loading capacity.
- Supports additional Duran bottles without the need for baskets.
- Stainless steel construction.
Specifications
Load Support Plate | |
Stainless Steel | |
1 Ea. |