missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Locator 6 Rack and Box System

Economically store up to 6,000 samples in the Locator 6 rack and box system that makes sample retrieval simple and consumes minimum liquid nitrogen.

Brand:  Thermo Scientific™ CY509113

5632.03 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 15529975

  • £8001.27 / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Manual Fill LN2 Dewar
184 L
Level Monitor
185 Days
95.3 cm
8.5 in.
21.5 cm
110/220 V
50/60 Hz
6000
72.6 kg
10 Boxes
6.2 cu. ft.
Vacuum
Locator 6
37.5 in.
26 in.
66 cm
Ultrasonic Level
110 - 220V 50/60 Hz
0.99L
160 lb.
Boxes (60/System)
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Locator 6 Rack and Box System >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.