Learn More
Eppendorf™ Locking Clip
Prevent spring action of tip cones with these locking clips
Brand: Eppendorf™ 3122613002
7.30 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory for Eppendorf Xplorer™/Xplorer plus
- Locking clip
- Available for 8- and 12-channel pipettes
Specifications
Locking Clip | |
Preventing spring action of tip cones | |
12-channel pipette | |
1 Each |