missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Locking Test Tube Rack

Test tube locking rack (holds 50 tubes). The Fisherbrand Locking Test Tube Rack has silicon gripping feature securely holds tubes in place.

Brand:  Fisherbrand™ 14955035

Product Code. 15916192

  • £8.10 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Holds 13 and 16mm test tubes
  • Securely grips up to 50 tubes
  • Made from Acrylonitrile butadiene styrene
  • Blue
Specifications

Specifications

Locking Test Tube Rack
Blue
ABS
Holding and pouring out the contents of tubes
10 x 5
Tubes (13 to 16 mm)
50
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.