missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Delivery Adapter, Long, Bent
For feeding distillate from condensers into receiving flasks. Lenz Laborglasinstrumente™ Delivery Adapters, Long, Bent are manufactured from durable DURAN™ glass tubing.
Brand: Lenz Laborglasinstrumente™ 05263004
Description
- With NS socket
- Long bent tube with angle of 105°
- 12mm diameter
Specifications
Long Bent Delivery Adapter | |
Duran Glass | |
NS 29/32 |