Learn More
Lenz Laborglasintrumente Long Ground Sockets
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100634
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10234191
  • £207.00 / Pack of 10
Estimated Shipment: 02-09-2024
to see stock.
Add to basket
Description
Description
  • Conical ground sockets
  • Made of DURAN™ tubing
Specifications
Specifications
10/Pk.
2.3 mm
Borosilicate Glass (3.3)
NS 34/45
40 mm