Learn More
Hamilton™ Long Life Instrument Syringes
Product Code. 10420465
Click to view available options
Volume (Metric):
25 μL
50 μL
100 μL
150 μL
250 μL
500 μL
1 mL
5 mL
Unit Size:
Each
Product Code. 10420465
Brand: Hamilton™ 825035
Long life syringes were developed for extreme conditions that include harsh chemicals as well as abrasive particulate matter.
  • Plunger tip made from UHMWPE (Ultra High Molecular Weight Poly Ethylene) which is harder than PTFE to better resist scratching
  • Plunger rod made from 316 stainless steel which offers exceptional chemical resistance
Specifications
Material Borosilicate Glass, PTFE, UHMWPE
Diameter (Metric) Outer 9.02 mm
Quantity 1 Ea.
Volume (Metric) 500 μL
Connection Type Luer Lock