missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ X-VIVO™ 10 Hematopoietic Serum-Free Culture Media
For adoptive immunotherapy and cellular therapy research
£261.49
Specifications
Content And Storage | 2° to 8°C |
---|---|
Product Type | Stem Cell Serum Free Media (SFM) |
Product Code | Brand | Without Additives | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Without Additives | Quantity | Price | Quantity & Availability | ||||
11675120
|
Lonza™
BE04-380Q |
1L |
£261.49
1 L |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Does not contain any lymphokines required for activation of human lymphocytes
- Intended for in vitro procedures
- Device Master Records (DMRs) for these products are held in an FDA Master File
- Chemically defined media optimized for specific cell types
- For further manufacturing or laboratory use
- Designed to support the generation of lymphokine activated killer (LAK) cells in a serum-free environment
- Limited by Federal (USA) law to investigational use
- These products are for research use only
- Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
Specifications
2° to 8°C | |
Stem Cell Serum Free Media (SFM) |