missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ IsoGel™ IEF Plates
Precast agarose IEF plates provide rapid and reproducible isoelectric focusing of proteins up to 2000kDa
£700.00
Specifications
Product Type | Gel Plate |
---|---|
Separation Type | Isoelectric Focusing |
Form | Solid |
For Use With (Application) | Isoelectric focusing, Antibody separation and analysis |
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
11507995
|
Lonza™
56015 |
3 to 10 |
£700.00
Pack of 6 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
11517995
|
Lonza™
56018 |
6 to 10.5 |
£700.00
Pack of 6 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
12890125
|
Lonza™
56024 |
7 to 11 |
£700.00
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
Description
- Specifically designed for antibody analysis of
- Each gel is supported on GelBond™ film for easy handling and dimensional stability
- Gel size is compatible with most IEF chambers
- 125 x 100mm
Isoelectric focusing, antibody separation and analysis
Order Info
Accessories packs (Cat. No. BMA56014 and BMA56010) and blotting paper (BMA56007) available separately.
Specifications
Gel Plate | |
Isoelectric Focusing | |
Solid | |
Isoelectric focusing, Antibody separation and analysis |