missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Latitude™ HT Precast Gels

Ready-to-use gels for high-throughput DNA electrophoresis. Latitude™ HT Precast Gels are precision cast for accurate, reproducible resolution of DNA fragments for 8bp to 10kb with our SeaKem™ or NuSieve™ Agarose.

£628.00 - £641.00

Specifications

Concentration 1%
Product Type Precast Gel
Dnase Free Yes
For Use With (Application) High throughput DNA electrophoresis
Material Agarose
View More Specs

Alert:

Ethidium bromide is a powerful mutagen; gloves should be worn when opening pouches and handling gels and any solutions containing ethidium bromide


Products 2
Product Code Brand Buffer Wells Separation Range Formulation Price Quantity & Availability  
Product Code Brand Buffer Wells Separation Range Formulation Price Quantity & Availability  
11557875
View Documents
Lonza™
57224
TBE 2 x 50-well 400 to ≥10,000 bp 1% SeaKem™ LE Plus Agarose, 1X TAE, prestained with 0.5μg/mL of Ethidium Bromide
£628.00
Each
Please to purchase this item. Need a web account? Register with us today!
11517885
View Documents
Lonza™
57234
TBE 4 x 50-well 300 to ≥8000 bp 1% SeaKem™ LE Plus Agarose, 1X TBE, prestained with 0.5μg/mL of Ethidium Bromide
£641.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Provide rapid, reproducible DNA resolution of 8 bp to ≥10kb
  • Manufactured from high-quality molecular biology grade SeaKem™ and NuSieve™ Brand Agarose
  • Quality and performance tested
  • Fit most large submerged electrophoresis chambers
  • Compatible with 8- and 12-tip multichannel pipets and microtiter plate spacing
  • Precision cast in accordance with ISO 9001 standards
  • Come in 100- and 200-well formats
  • Prestained with 0.5μg/mL of ethidium bromide
  • Permit runs to be completed in 30 minutes
  • Allow up to 200 samples to be run on a single precast gel

Specifications

Specifications

1%
Yes
Agarose
Yes
0.25 in.
10 to 12 μL
14 cm
Precast Gel
High throughput DNA electrophoresis
5 Gels
18° to 26°C
0.65 cm
24 cm
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 6 to 12 Months

SDS
Documents

Documents

Product Certifications