missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NuSieve™ 3:1 Agarose, Lonza™
Click to view available options
Quantity:
125 g
Specifications
CAS | 9012-36-6 |
Quantity | 125 g |
Grade | Molecular Biology |
Chemical Name or Material | Agarose, NuSieve 3:1 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'