missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Reliant™ Precast Gels

For rapid and reproducible resolution of DNA sizes from 8bp-10kb - Lonza™ Reliant™ Precast Gels are ideal for analysis of PCR, restriction digest fragments or most cloning needs.

Brand:  Lonza™ 54927

 View more versions of this product

Product Code. 11557825

  • £384.00 / Pack of 20
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket

Alert:

Ethidium bromide is a powerful mutagen. Gloves should be worn when opening pouches and handling gel solutions containing ethidium bromide.

This item is not returnable. View return policy

Description

Description

  • Manufactured with high-quality SeaKem™ Gold and NuSieve™ 3:1 Plus brand agaroses
  • Easy to handle
  • Fit most common mini submerged electrophoresis systems
  • Available with 0.5μg/mL of ethidium bromide for ease of visualizing DNA bands during and after the run
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TBE
4%
8 to ≥1000 bp
Yes
DNA separation by gel electrophoresis
20 Gels
18° to 26°C
0.5 cm
3.7 in.
2.37 in.
Clear
Precast Gel
8-well
4% NuSieve 3:1 Plus Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide
Agarose
Yes
0.19 in.
≤15 μL
9.5 cm
6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers