missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Reliant™ UV Transparent Gel System Tray
With an open-ended design for clear, straight band patterns
Brand: Lonza™ 54946
Additional Details : Weight : 0.45000kg
Description
- Solid, heavy acrylic construction eliminates need for tape when recommended minimum buffer levels are used
- Remove gel from package tray, place in reusable tray, load and run
- Portrait layout
- 2.6 x 3.9 in. (6.7 x 9.9cm)
Specifications
Transparent | |
Gel System Tray | |
Acrylic |
Electrophoresis System | |
Portrait |