Learn More
Lonza™ SYPRO™ Ruby Protein Blot Stain
Offers high-sensitivity detection on PVDF and nitrocellulose membranes
Brand: Lonza™ 50565
Additional Details : Weight : 1.00000kg
Description
Does not interfere with subsequent immunodetection.
Product licensed by Cambrex from Molecular Probes, Inc. For research and quality control use only.
Specifications
Ruby | |
Liquid | |
200 mL |
Protein Stain | |
Fluorogenic, chemiluminescent and colorimetric detection techniques |