missing translation for 'onlineSavingsMsg'
Learn More

IKA™ Loopster Basic
GREENER_CHOICE

Brand:  IKA™ 0004015000

607.37 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 13497659

  • £827.00 / Each
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Order Info

Note: Attachment discs need to be ordered separately.

Specifications

Specifications

0 to 20 rpm
100/240 V
1 A
50/60 Hz
300 mm
80%
100/240 V, 24 VDC, 50/60Hz, 1A
4°C to 50°C
IP 21 DIN EN 60529
300 mm
Loopster Basic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers