missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Losartan (D4 Carboxylic Acid), MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Losartan D4 Carboxylic Acid (E-3174 D4) is the deuterium labeled Losartan(EXP-3174), which is an angiotensin II receptor antagonist.
Specifications
Specifications
| Chemical Name or Material | Losartan (D4 Carboxylic Acid) |
| CAS | 1246820-62-1 |
| Purity Grade Notes | Research |
| Physical Form | Powder |
| Molecular Formula | C22H17D4ClN6O2 |
| Color | Off-White |
| Synonym | E-3174 D4 EXP-3174 D4 |
| SMILES | ClC1=C(C(O)=O)N(CC2=C([2H])C([2H])=C(C3=CC=CC=C3C4=NN=NN4)C([2H])=C2[2H])C(CCCC)=N1 |
| Industry Type | Metabolism-protein/nucleotide metabolism |
| Molecular Weight (g/mol) | 440.92 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction