missing translation for 'onlineSavingsMsg'
Learn More

Losartan potassium, Tocris Bioscience™

Product Code. 18752634
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective, non-peptide AT1 antagonist

Selective non-peptide angiotensin AT1 receptor antagonist. Inhibits the contractile effects of angiotensin II on rabbit aorta and jugular vein (pA2 = 8.27). Orally active antihypertensive agent.

Specifications

CAS 124750-99-8
Molecular Formula C22H25ClKN6O
Quantity 50 mg
Synonym 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)-[1, 1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-methanol potassium salt
InChI Key GKVBJVGUWMACMS-UHFFFAOYSA-N
SMILES [HH].CCCCC1=NC(=C(N1CC2=CC=C(C=C2)C3=CC=CC=C3C4=NNN=N4)CO)Cl.[K]
IUPAC Name [2-butyl-5-chloro-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol;molecular hydrogen;potassium
Molecular Weight (g/mol) 464.031
PubChem CID 131674082
Formula Weight 461
Percent Purity >99%
Chemical Name or Material Losartan potassium
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.