missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine HR

Product Code. 10224982
Click to view available options
Range:
10 to 300mg/L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10224982

Brand: Lovibond™ 147030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • Aqua Comparator Housing
  • Aqua Comparator Disc
  • Acidifying GP Tablets (100 pack)
  • Chlorine HR (KI) Tablets (100 pack)
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying Case

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Accuracy ±5%
For Use With (Application) Routine water analysis
Includes Aqua comparator™ housing, aqua comparator™ disc, acidifying GP tablets, chlorine HR (KI) tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
No. of Tests 100
Range 10 to 300mg/L
Type Chlorine Free and Total Test Kit
compliance-icons
Product Identifier
  • COMPARATOR LOVIBOND 147030 CHLORINE HR
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : citric acid,adipic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.