missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ COD Tube Tests, HR, Mercury Free
Click to view available options
Range:
0 to 15000mg/L
Unit Size:
Pack of 25
Description
- For use in SC400™ Colorimeters and Aqua Comparator™ Test Kits
- Also suitable for use in Hach™ instruments and test kits
- Easy-to-open, sealed vials eliminate risk of cross-contamination
- Extended shelf life of up to five years
For Municipal Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted test method
Specifications
Specifications
| Certifications/Compliance | US EPA |
| For Use With (Application) | Municipal wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | SC400 colorimeters and Aqua comparator™ test kits, Hach™ instruments |
| No. of Tests | 25 |
| Range | 0 to 15000mg/L |
| Test Method | Dichromate/H2SO4 |
Sicurezza e movimentazione
Identificativo prodotto
- COD / CSB Mercury Free, 0-15000 mg/l
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Germ cell mutagenicity Category 1B
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH208-Contains potassium dichromate. May produce an allergic reaction.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : sulphuric acid, disilver(1+) sulfate, potassium dichromate
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto