missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Color of Varnish (Gardner Color) Discs
£355.00
Specifications
Certifications/Compliance | ASTM D-1544, AOCS Td 1a |
---|---|
Type | Color of Varnish l Disc |
For Use With (Equipment) | Varnish comparator kits 705-V2, 705-VA2 or 705-VA4 |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
15833032
|
Lovibond™
243030 |
1 to 9 |
£355.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15843032
|
Lovibond™
243040 |
9 to 18 |
£355.00
Each |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ASTM D-1544, AOCS Td 1a | |
Color of Varnish l Disc | |
Varnish comparator kits 705-V2, 705-VA2 or 705-VA4 |