missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ DPD No. 3 HR Tablets

Product Code. 15163058
Click to view available options
Range:
0.01 to 10ppm
Unit Size:
100 tablets
This item is not returnable. View return policy

Product Code. 15163058

Brand: Lovibond™ 511590BT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color White
Composition Potassium Iodide <30%, non-hazardous components <70%
pH Range 6.3
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) SC450 chlorine 2 and 3 colorimeters, MC500 colorimeter, SP600 spectrophotometer, AquaPRO3 and PRO6 Pool photometers
Range 0.01 to 10ppm
Storage Requirements 20°C
Test Method DPD
Type DPD No. 3 HR Tablet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.