missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ DPD No. 4 Tablets

Convenient blister packaging allows user to push tablet through foil directly into sample

Brand:  Lovibond™ 511220BT

Product Code. 10515730

  • £34.30 / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Convenient blister packaging allows user to push tablet through foil directly into sample

  • Each tablet hermetically sealed in individual aluminum foil pocket that protects it from environmental conditions for longer shelf life
  • For use with MC500™ Multi-Parameter Handheld Colorimeter (Cat. No. 15-398-470) and SP600 Spectrophotometer (15-398-320)
  • Tests for active oxygen in water samples
  • Range: 0.1 to 10mg/L

Specifications

Specifications

White
6.2
Water analysis
0.1 to 10mg/L
DPD
Potassium Iodide 1 to 10%, N,N-diethylbenzene-1,4-diammonium Sulphate 0.1 to 1%, non-hazardous components >90%
Tablet
MC500 colorimeter and SP600 spectrophotometer
20°C
DPD No. 4 Tablet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers