Learn More
Lovibond™ Halogen Lamp for SP600 Spectrophotometer
Click to view available options
Type:
Replacement Tungsten Halogen Lamp Assembly
Unit Size:
Each
Description
Replacement tungsten halogen lamp assembly (6V,10w) for SP600 Spectrophotometer.
Replacement tungsten halogen lamp assembly (6V,10w) for SP600 Spectrophotometer.
Specifications
Specifications
| Type | Replacement Tungsten Halogen Lamp Assembly |
| For Use With (Equipment) | SP600 spectrophotometer |