Learn More
Lovibond™ Infrared Lab Turbidimeter
Designed for quick and easy turbidity measurement in the laboratory
Brand:  Lovibond™ 194000
Product Code. 10247642
  • £1655.00 / Each
Explore more special offers
Includes: All items necessary for testing including turbidimeter, four sample cells, calibration set, operator's manual, and carrying case. (Batteries sold separately.)
Description
Description
  • Auto-ranging function automatically selects the correct range for turbidity measurement from 0.01 to 1100 NTU
  • Data-logging feature enables user to store up to 1,000 data sets with the date, time, and registration number
  • Instrument is power efficient; operates on seven NiCd rechargeable batteries (sold separately) or wall plug adapter
Specifications
Specifications
Infrared Lab Turbidity Meter
Color Graphic Display
12 mL
±2%, ±5%
0.01 to 1100 NTU
10.45 in.
7.6 in.
2.76 in.
NiCd rechargeable
50/60 Hz
CE, ISO
5°C to 40°C
1 kg
Infrared LED
Nephelometric
For quick and easy turbidity measurement in the laboratory
0.01, 0.1, 1 NTU
10.45 x 7.6 x 2.76 in. (26.5 x 19.3 x 7 cm)
26.5 cm
19.3 cm
7 cm
100/230 V 50/60 Hz
100/230 V
41°F to 104°F
2.2 lb.