missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MD600 Handheld Water Quality Colorimeter

Rugged, waterproof multi-parameter colorimeters for laboratory and field testing

Brand:  Lovibond™ 214020

Code : NEW

Additional Details : Weight : 2.16000kg

Product Code. 15348314

  • £1050.00 / Each
Estimated Shipment: 08-01-2025
to see stock.

Includes:

Instrument in carrying case, three 24mm sample cells, three 16mm sample cells, one 13mm tube adapter, one 16mm tube adapter, four AA batteries, an operator's manual and a carrying case


Description

Description

Rugged, waterproof multi-parameter colorimeters for laboratory and field testing

Combine the mobility of a portable photometer with the characteristics of a modern laboratory photometer. The waterproof design makes them ideal for a field instrument, and they have the functionality and features to be used as laboratory photometers. The units cover all the important parameters for water analysis from aluminum to Zinc and have up to 120 methods pre-programmed. Compatible with Tablet, Liquid and Powder reagents and supplied with vial adaptors for 13mm and 16mm tubes, the photometers give flexibility of choice for many tests. In addition to the pre-programmed methods, up to 10 user defined calibration curves can be programmed if required.

A large graphic display aids analysis by providing on screen method prompts, information regarding test measurement range and reagent type, and automatic countdown timers for accurate reaction periods. The internal memory is capable of storing up to 1000 results with date, time and sample ID. These results can be reviewed at any time and can be downloaded to a PC via an additional Infra Red module (IRiM). With 6 LED light sources, the photometers can measure at peak absorbances for many different methods, ensuring result accuracy and reliability for many applications.

  • Backlit display enables easy readings
  • Interfaces: Infra red interface for test data transfer (via IRiM); RJ45 socket for internal updates
  • Six wavelength photo detector array
  • Light-emitting diodes and photosensor-intensifier with automatic wavelength selection (430, 530, 560, 580, 610 and 660nm)
  • Acid- and solvent-resistant, touch-sensitive keypad with audible feedback
  • Supports 70 pre-programmed methods and 20 user-defined methods (10 with calibration point input, 10 with polynominal input)
  • Internal memory stores up to 1000 data sets which can be transferred to a computer
  • Automatically turns off 20 minutes after the last key press with an audible signal
  • Ambient conditions: 5 to 40°C; 30-90% relative humidity (noncondensing)
  • User interface in English, German, French, Spanish, Italian, Portuguese and Polish
Dimensions: 8.27L × 3.7W × 1.8 in.H (21 × 9.39 × 4.57cm).
Weight: 1 lb. (450g).
  • Power supply: four AA batteries (26 hr. operation or 3500 tests)
  • Compliance

    CE marked

    Services

    Two years

    Specifications

    Specifications

    0.02
    CE
    8.3 x 3.7 x 1.8 in. (21 x 9.3 x 4.5 cm)
    English, German, French, Spanish, Italian, Portuguese, Polish, Bahasa Indonesian
    1.5 V
    Laboratory and field testing
    4.5 cm
    21 cm
    2 Years
    0.454 kg
    9.3 cm
    Yes
    LCD
    IP68
    Multi-parameter Colorimeter
    430, 530, 560, 580, 610, 660 nm
    1.8 in.
    8.3 in.
    0.005 A
    1 lb.
    3.7 in.
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers