Learn More
Lovibond™ Powder Pack™ Reagents for Ortho (Free) Phosphate
Pre-measured, unit dose in a convenient foil pack
Brand:  Lovibond™ 531550
Product Code. 15311675
  • £47.75 / Pack of 100
Explore more special offers
Description
Description
  • Easy to handle
  • Easy to open
  • Easy to dispense and pour
  • Specifically formulated for use in Hach™ and other manufacturer's test kits
Specifications
Specifications
Powder Pack Reagents
Orbeco and Hach colorimeters and spectrophotometers
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • vario sulphate 4
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : citric acid, bariumchloride dihydrate