missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Total Chlorine Powder Packs™
Premeasured unit dose DPD reagents for water quality testing
£22.10 - £194.00
Specifications
Certifications/Compliance | US EPA |
---|---|
Format | Powder |
For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
For Use With (Equipment) | SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer |
Type | DPD Total Chlorine Reagent |
Product Code | Brand | Quantity | Range | Volume (English) | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Range | Volume (English) | Volume (Metric) | Price | Quantity & Availability | |||
15351645
|
Lovibond™
530080 |
100 Pack | 0 to 3.5 mg/L | 0.16 oz. | 5 mL |
£22.30
Pack of 100 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||
15361645
|
Lovibond™
530083 |
1000 Pack | 0 to 3.5 mg/L | 0.16 oz. | 5 mL |
£131.00
Pack of 1000 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||
15341655
|
Lovibond™
530123 |
1000 Pack | 0 to 2 mg/L | 0.33 oz. | 10 mL |
£173.00
Pack of 1000 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||
10402345
|
Lovibond™
530120 |
100 Pack | 0 to 2 mg/L | 0.33 oz. | 10 mL |
£22.10
Pack of 100 |
Estimated Shipment: 14-03-2025
Log in to see stock. |
|||
15351655
|
Lovibond™
530130 |
100 Pack | 0 to 5 mg/L | 0.84 oz. | 25 mL |
£25.55
Pack of 100 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||
15361655
|
Lovibond™
530133 |
1000 Pack | 0 to 5 mg/L | 0.84 oz. | 25 mL |
£194.00
Pack of 1000 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||
Description
Premeasured unit dose DPD reagents for water quality testing
- For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
- Also suitable for use in Hach™ instruments and test kits
- Easy-to-open, sealed foil packets eliminate risk of cross-contamination
- Available for 5, 10, and 25mL sample sizes
- Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
US EPA | |
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
DPD Total Chlorine Reagent |
Powder | |
SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer |
Safety and Handling
missing translation for 'shelfLife' : 5 Years
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lovibond™ Total Chlorine Powder Packs™