Learn More
Lovibond™ Powder Packs™: for Tube Test; Nitrite; Low Range; Nitrite 3; 25mL
Premeasured unit dose reagent for water quality testing
Brand: Lovibond™ 530970
Code : NEW
Additional Details : Weight : 0.30000kg Transport : UN number : 3260 Chem class : 8 Pack group : III
Description
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Orbeco-Hellige and Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
US EPA | |
Potassium Dihydrogenorthophosphate 75 to 85%, Sodium Sulfanilite Dihydrate 5 to 10%, Potassium Disulfate 1 to 10%, non-hazardous components 1 to 5% | |
25 mL | |
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
0 to 0.3mg/L | |
0.85 oz. |
Light Beige | |
3.1 | |
Powder | |
Hach™ and other manufacturers test kits, colorimeters and spectrophotometers | |
Nitrite 3 Reagent |
Safety and Handling
- POWDER VARIO NITRI3 25 ML
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : dipotassium disulphate